Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enterobacter aerogenes Antibody (57/82), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308869G
Description
Enterobacter aerogenes Monoclonal antibody specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
Enterobacter aerogenes | |
Monoclonal | |
DyLight 488 | |
Enterobacter aerogenes, Klebsiella mobilis | |
A crude sonicate of Enterobacter aerogenes | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG κ |
ELISA, Immunofluorescence | |
57/82 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction