Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enterobacter Cloacae Antibody (61/212), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062544V
Description
Enterobacter Cloacae Monoclonal specifically detects Enterobacter Cloacae in Bacteria samples. It is validated for ELISA.Specifications
Enterobacter Cloacae | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
This product recognizes Enterobacter cloacae, a gram-negative bacillus sometimes associated with urinary and respiratory tract infections in humans. | |
Store at 4C in the dark. | |
IgG2a |
ELISA | |
61/212 | |
ELISA | |
Bacillus cloacae, Bacterium cloacae, Cloaca cloacae | |
Enterobacter cloacae NCTC 10005 | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction