Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enterobacteriaciae ECA Monoclonal Antibody (1055)
GREENER_CHOICE

Catalog No. PIMA533256
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533256 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533256 Supplier Invitrogen™ Supplier No. MA533256
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For ELISA sandwich assay, employ MA5-33256 as biotin conjugate.

Enterobacterial common antigen (ECA) is a family-specific surface antigen shared by all members of the Enterobacteriaceae and is restricted to this family. ECA is found in freshly isolated wild-type strains as well as in laboratory strains like Escherichia coli K-12. The family specificity of ECA can be used for taxonomic and diagnostic purposes. ECA is located in the outer leaflet of the outer membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterobacteriaciae ECA
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1055
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Enterobacteria
Host Species Mouse
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.