Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enterococcus Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA173120
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173120 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173120 Supplier Invitrogen™ Supplier No. PA173120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73120 antibody reacts with Enterococcus species and may react with related microorganisms. PA1-73120 has been successfully used in immunofluorescent and ELISA applications. The PA1-73120 antibody was raised against Enterococcus faecium whole cells. For longer term storage, aliquot and freeze.

Enterococci form part of the lactic acid bacteria family and are the third most important bacterial genus in hospital infections, especially Enterococcus (E.) faecium. In food they can spoil processed meats but they are equally important for ripening and aroma development of certain traditional cheeses and sausages and are used as human probiotics. However, they possesses a broad spectrum antibiotic resistance which has important implications to human health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterococcus
Applications Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias E. faecium; Enterococci
Host Species Rabbit
Immunogen Enterococcus faecium whole cells.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.