Learn More
Invitrogen™ Enterococcus Polyclonal Antibody, Biotin

Description
The PA1-73123 antibody reacts with Enterococcus species. PA1-73123 has been successfully used in immunofluorescence and ELISA applications. The PA1-73123 antibody was raised against Enterococcus faecium whole cells. Recommended short term storage, up to 6 months, is liquid at 4ºC. For longer term storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Enterococcus |
| Applications | ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | E. faecium; Enterococci |
| Host Species | Rabbit |
| Immunogen | Enterococcus faecium whole cells. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.