Learn More
Invitrogen™ Enterococcus Polyclonal Antibody, FITC

Description
The PA1-73121 antibody reacts with Enterococcus species. PA1-73121 has been successfully used in Immunofluorescence applications. Aceton fixation of the antigen source is recommended prior to staining. The PA1-73121 antibody was raised against Enterococcus faecium whole cells. Recommended short term (6 months) storage is liquid at 4°C under subdued light. For longer term storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Enterococcus |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5.0 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | E. faecium; Enterococci |
| Host Species | Rabbit |
| Immunogen | Enterococcus faecium whole cells. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.