Learn More
Invitrogen™ Enterococcus Polyclonal Antibody, HRP

Description
The PA1-73122 antibody reacts with Enterococcus species. PA1-73122 has been successfully used in ELISA applications. The PA1-73122 antibody was raised against Enterococcus faecium whole cells.
Specifications
Specifications
| Antigen | Enterococcus |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1-2 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 10mg/mL BSA and 0.002% thimerosal |
| Gene Alias | E. faecium; Enterococci |
| Host Species | Rabbit |
| Immunogen | Enterococcus faecium whole cells. |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.