Learn More
Enterokinase Cleavage Site Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA130443
Description
PA1-30443 detects ECS from rabbit samples. PA1-30443 has been successfully used in immunocytochemistry, immunohistochemistry, Western blot applications. The PA1-30443 immunogen is: Rabbits were immunized with ECS enterokinase cleavage site (xxxDDDDK). Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.

Specifications
Enterokinase Cleavage Site | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Purified | |
RUO | |
Rabbit | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
1 mg/mL | |
ECS; ECS Cleavage Site | |
Rabbits were immunized with ECS enterokinase cleavage site (xxxDDDDK). Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
100 μg | |
Primary | |
4° C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.