Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterokinase Mouse anti-Bovine, Porcine, Clone: 5E3C8, GenScript™

Catalog No. 50273031
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-031 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-273-031 Supplier Genscript Corporation Supplier No. A0088940
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/mL. Enterokinase is an intestinal enzyme responsible for initiating activation of pancreatic proteolytic proenzymes. Mouse Anti-Enterokinase Monoclonal Antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified recombinant EK protein (bovine EK, aa848-1035, light chain, 93% homologous with human EK).Mouse Anti-Enterokinase Monoclonal Antibody recognizes EK in Western blots and ELISAs.

Enterokinase Monoclonal antibody specifically detects Enterokinase in Bovine, Porcine samples. It is validated for ELISA, Western Blot

Specifications

Antigen Enterokinase
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5E3C8
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene TMPRSS15
Gene Accession No. P98072, P98074
Gene Alias enterokinase; enterokinase catalytic subunit; enteropeptidase; Enteropeptidase catalytic light chain; Enteropeptidase non-catalytic heavy chain; Entk; protease, serine, 7 (enterokinase); PRSS7; Serine protease 7; Tmprss15; Transmembrane protease serine 15; transmembrane protease, serine 15
Host Species Mouse
Immunogen Purified recombinant light chain of bovine EK protein
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282009, 397152
Target Species Bovine, Pig
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.