Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enteropeptidase/Enterokinase Antibody (MM0259- 8S12), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212411AF488
Description
Enteropeptidase/Enterokinase Monoclonal specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot.Specifications
Enteropeptidase/Enterokinase | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TMPRSS15 | |
Human recombinant Enteropeptidase/Enterokinase | |
0.1 mL | |
Primary | |
Detects human Enteropeptidase/Enterokinase heavy chain | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
MM0259- 8S12 | |
Western Blot | |
EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
Mouse | |
Protein G purified | |
ECS Epitope Tags (DDDDK), Epitope Tags, Immunology, Innate Immunity | |
5651 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction