Learn More
Liofilchem™ EnteroPluri-Test
SDP
Catalog No. 22778079 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
10 Pk.
25 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-778-079 25 Pk.
22-778-080 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-778-079 Supplier Liofilchem™ Supplier No. 78619U
Add to Cart
Add to Cart
This item is not returnable. View return policy
EnteroPluri-Test is a 12-sector system containing special culture media for the identification of Enterobacteriaceae and other Gram-negative, oxidase negative bacteria.
  • Allows the simultaneous inoculation of all media present in the sectors and the execution of 15 biochemical reactions
  • Microorganism is identified evaluating the color change of different culture media after 18 to 24 hr incubation at 36 ±1°C and by code number obtained from interpretation of the biochemical reactions
  • Used for identification of Gram-negative, oxidase negative bacteria isolated on selective culture media for Enterobacteriaceae, such as: MacConkey Agar (MCA), Eosin Methylene Blue Agar (EMBA), Salmonella and Shigella Agar (SSA), Hektoen Enteric Agar (HEEA), or non-selective culture media
  • Tests come individually packed
Specifications
Disposable Single-use
For Use With (Application) Identification of Gram-negative or Oxidase Negative Bacteria
Quantity 25 Pk.
Non-hazardous' according to current legislation, but falls within specific field of application where a safety data sheet must be supplied, as product can cause phenomena of sensitization in sensitive subjects if in contact with skin.