Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enterovirus 71 3D Monoclonal Antibody (4)
GREENER_CHOICE

Catalog No. PIMA535903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535903 Supplier Invitrogen™ Supplier No. MA535903
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enterovirus 71 3D Monoclonal Antibody for Western Blot, ICC/IF, IP

EV71 is a positive-sense single-stranded RNA virus of the Picornaviridae family. It is known to cause hand, foot, and mouth disease and various neurological pathological conditions, such as aseptic meningitis and ataxia. EV71 is shown to utilize host UDP-glucose glycoprotein glucosyltransferase 1 (UGGT1) to enhance viral replication and virulence. It forms replication complexes (RCs) on cellular membranes that contain a mix of host and viral proteins to facilitate viral replication. During infection viral RNA replication occurs on the cytoplasmic surfaces of single-membrane vesicles derived from the endoplasmic reticulum, and the membranes can serve to accelerate RC assembly during positive-strand genomic RNA replication. It has been shown that 3D polymerase, an RNA-dependent RNA polymerase that is encoded in the P3 viral genome region, plays a key role in EVA71 replication. Neural progenitor cells are shown to be prone to EV71 infection and that these stem cells can support the active replication of this virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus 71 3D
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative; pH 7
Gene Alias Enterovirus
Host Species Mouse
Immunogen The immunogen used to generate this antibody corresponds to EV71 3D.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.