Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterovirus 71 3D Mouse anti-Virus, Unconjugated, Clone: 4, MilliporeSigma™

Catalog No. MABF22352MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22352MI 25 μg
MABF22351MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22352MI Supplier MilliporeSigma Supplier No. MABF223525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enterovirus 71 3D Monoclonal antibody specifically detects Enterovirus 71 3D in Virus samples. It is validated for Immunofluorescence, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus 71 3D
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4
Conjugate Unconjugated
Gene Alias EV71;EV71 3D polymerase;Human rhinovirus A71
Gene Symbols CN12_48981gpPO
Host Species Mouse
Immunogen Recombinant 3D polymerase from Enterovirus 71.
Molecular Weight of Antigen ∼52 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 4 is a mouse monoclonal antibody that specifically detects Enterovirus 71 3D polymerase in EV71 infected cells.
Target Species Virus
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.