Learn More
Description
Specifically detects Enterovirus 71 in Monkey, Mouse, Virus samples, and it is validated for ELISA, Immunocytochemistry, Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | Enterovirus 71 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 6.15mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000-1:20000, ELISA 1:100 - 1:2000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 |
| Gene Alias | EV71 |
| Host Species | Rabbit |
| Immunogen | The immunogen used to generate this antibody corresponds to EV71 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
