Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enterovirus 71 Monoclonal Antibody (10F0)
GREENER_CHOICE

Catalog No. PIMA183142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183142 Supplier Invitrogen™ Supplier No. MA183142
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody cross-reacts with Coxsackie A16. Mouse Anti Enterovirus 71 antibody clone 422-8D-4C-4D (10F0), recognizes Enterovirus 71.

Entervirus 71 is a potentially fatal virus first isolated in 1969. Enterovirus 71 has been found in the spinal cord and medulla and can produce a range of symptoms from rashes to meningitis and ecephalitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus 71
Applications Functional Assay, Immunocytochemistry
Classification Monoclonal
Clone 10F0
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with no preservative
Gene Alias EV71
Host Species Mouse
Immunogen Enterovirus 71 strain BrCr.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.