Learn More
Invitrogen™ Enterovirus 71 VP1 Monoclonal Antibody (3656)

Description
Non-reactive with: Coxsackie A9, Coxsackie B2, ECHO 30, EV70 and Polio, type 2.
Specifications
Specifications
| Antigen | Enterovirus 71 VP1 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3656 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | Enterovirus; EV71 |
| Host Species | Mouse |
| Purification Method | Protein A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.