Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Enterovirus 71 VP1 Monoclonal Antibody (GT9512)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA535907
Description
Enterovirus 71 VP1 Monoclonal Antibody for Western Blot, ELISA
Entervirus 71 is a potentially fatal virus first isolated in 1969. Enterovirus 71 has been found in the spinal cord and medulla and can produce a range of symptoms from rashes to meningitis and ecephalitis.
Specifications
Enterovirus 71 VP1 | |
Monoclonal | |
1.11 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Western Blot | |
GT9512 | |
Unconjugated | |
Enterovirus; EV71 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of EV71 virus VP1 protein. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction