Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Enterovirus 71 VP1 Recombinant Rabbit Monoclonal Antibody (HL1929)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547224
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Entervirus 71 is a potentially fatal virus first isolated in 1969. Enterovirus 71 has been found in the spinal cord and medulla and can produce a range of symptoms from rashes to meningitis and ecephalitis.
Specifications
Enterovirus 71 VP1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA, Western Blot | |
HL1929 | |
Unconjugated | |
Enterovirus; EV71 | |
Synthetic peptide of Enterovirus 71 VP1. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction