Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enterovirus 71 VP1 Recombinant Human Monoclonal Antibody (H136)

Catalog No. PIMA529968
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529968 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529968 Supplier Invitrogen™ Supplier No. MA529968

Human Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency. This antibody has specificity for EV Enterovirus 71 VP1.

Entervirus 71 is a potentially fatal virus first isolated in 1969. Enterovirus 71 has been found in the spinal cord and medulla and can produce a range of symptoms from rashes to meningitis and ecephalitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus 71 VP1
Applications Neutralization
Classification Recombinant Monoclonal
Clone H136
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Enterovirus; EV71
Host Species Human
Immunogen Enterovirus 71 VP1.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.