Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enterovirus D68 VP1 Monoclonal Antibody (GT162), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA535911

Description
Enterovirus D68 VP1 Monoclonal antibody specifically detects Enterovirus D68 VP1 in Virus samples. It is validated for Western Blot
Specifications
Enterovirus D68 VP1 | |
Monoclonal | |
2.8 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2b |
Western Blot | |
GT162 | |
Unconjugated | |
Enterovirus | |
Recombinant protein encompassing a sequence within the C-terminus region of Enterovirus D68 VP1 protein. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction