Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enterovirus D68 VP3 Monoclonal Antibody (GT1665)
GREENER_CHOICE

Catalog No. PIMA535913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535913 Supplier Invitrogen™ Supplier No. MA535913
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enterovirus D68 VP3 Monoclonal Antibody for Western Blot

Enterovirus D68 (EV-D68) has been recognized as an emerging pathogen after recent worldwide outbreaks of serious respiratory and neurological disease. Its capside proteins are VP1-VP4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus D68 VP3
Applications Western Blot
Classification Monoclonal
Clone GT1665
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Enterovirus
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of Enterovirus D68 VP3. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.