Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560308
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560308 Supplier Invitrogen™ Supplier No. PA560308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEEETVTTKH IHITQATETT TTRHKRTANP SKTIDLGAAA HYTGDKASPD QNASTHTPQS SVKTSVPSSK SSGDLVDLFD GTSQSTGGSA DLFGGFADFG S Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.

Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLINT1
Gene Accession No. Q14677, Q99KN9
Gene Alias AI642036; AW537616; C530049I24Rik; clathrin interacting protein localized in the trans-Golgi region; clathrin interactor 1; Clathrin interactor 1-like protein; Clathrin-interacting protein localized in the trans-Golgi region; Clint; Clint1; ENTH; ENTH domain containing protein; enthoprotin; Epn4; EPNR; epsin 4; epsin-4; epsinR; Epsin-related protein; F630206G17Rik; KIAA0171; mKIAA0171
Gene Symbols CLINT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ENTH. Recombinant protein control fragment (Product #RP-100124).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216705, 360515, 9685
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.