Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566581 Supplier Invitrogen™ Supplier No. PA566581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPILLKGTSDD DVPCPGYLFE EIAKISHESP GSSQCLLEYL LSRLHSSSGH GKLKVLKILL YLCSHGSSFF LLILKRNSAF IQEAAAFAGP PDPLHGNSLY QKVR Highest antigen sequence identity to the following orthologs - mouse 89%, rat 90%.

ENTHD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTHD2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEPSIN
Gene Accession No. Q96N21
Gene Alias 2410002I01Rik; AP-4 accessory protein; AP-4 complex accessory subunit tepsin; C17orf56; C19H17orf56; C9H17orf56; ENTH domain containing 2; ENTH domain-containing protein 2; Enthd2; Epsin for AP-4; protein C17orf56 homolog; RGD1307410; TEPSIN; TEPSIN adaptor related protein complex 4 accessory protein; TEPSIN, adaptor related protein complex 4 accessory protein; tetra-epsin
Gene Symbols TEPSIN
Host Species Rabbit
Immunogen Recombinant Human ENTHD2. Recombinant protein control fragment (Product #RP-105259).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146705
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.