Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTPD2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5110829

Catalog No. PIPA5110829


Only null left
Add to Cart

Description

Description

Immunogen sequence: SHTSMFIYKW PADKENDTGI VGQHSSCDVP GGGISSYADN PSGASQSLVG CLEQALQDVP KERHAGTPLY LGATAGMRLL NLTNPEASTS VLMAVTHTLT QYPFDFRGAR ILSGQEEGVF GWVTANYLLE NFIKYGW.

The protein encoded by this gene is the type 2 enzyme of the ecto-nucleoside triphosphate diphosphohydrolase family (E-NTPDase). E-NTPDases are a family of ecto-nucleosidases that hydrolyze 5'-triphosphates. This ecto-ATPase is an integral membrane protein. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ENTPD2
Polyclonal
Unconjugated
ENTPD2
CD39 antigen-like 1; CD39L1; Ecto-ATP diphosphohydrolase 2; ecto-ATPase 2; ecto-ATPDase 2; Ectonucleoside triphosphate diphosphohydrolase 2; ENTPD2; NTPDase 2; NTPDase2; NTPDase-2; testicular ecto-ATPase
Rabbit
Antigen affinity chromatography
RUO
954
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y5L3
ENTPD2
Recombinant Protein corresonding to Human ENTPD2. Recombinant protein control fragment (Product #RP-90917).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.