Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547625
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547625 Supplier Invitrogen™ Supplier No. PA547625
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 30% cross-reactivity with recombinant human CD39L3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

T-cadherin was initially identified as cadherin-type cell adhesion molecule expressed in various neuronal populations in a temporally and spatially restricted pattern during axon growth. T-cadherin is an atypical member of the cadherin family because it does not possess the typical transmembrane and cytoplasmic domains but is instead anchored to the plasma membrane by glycosylphosphatidylinositol (GPI) linkage. T-cadherin may play a role in malignant tumor development as loss of the chromosome locus containing the T-cadherin gene correlates with the development of a variety of cancers. Recently it has been shown that T-cadherin can act as a receptor for hexameric and high-molecular weight forms of adiponectin, suggesting that T-cadherin may also play a role in metabolic regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ENTPD3
Gene Accession No. 0
Gene Alias CD39 antigen-like 3; CD39L3; CD39-like 3; ecto-apyrase 3; Ecto-ATP diphosphohydrolase 3; ecto-ATPase 3; ecto-ATPDase 3; Ectonucleoside triphosphate diphosphohydrolase 3; ENTPD3; HB6; NTPDase 3; NTPDase3; NTPDase-3
Gene Symbols ENTPD3
Host Species Sheep
Immunogen CHO-derived recombinant mouse CD39L3/ENTPD3 Gln44-Pro485.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215446
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.