Learn More
ENTPD3 Polyclonal Antibody, Invitrogen™
Description
Specifications
Specifications
| Antigen | ENTPD3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | ENTPD3 |
| Gene Accession No. | O75355 |
| Gene Alias | CD39 antigen-like 3; CD39L3; CD39-like 3; ecto-apyrase 3; Ecto-ATP diphosphohydrolase 3; ecto-ATPase 3; ecto-ATPDase 3; Ectonucleoside triphosphate diphosphohydrolase 3; ENTPD3; HB6; NTPDase 3; NTPDase3; NTPDase-3 |
| Gene Symbols | ENTPD3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.