Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENTPD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522247
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (88%), Rat (86%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.
Specifications
ENTPD5 | |
Polyclonal | |
Unconjugated | |
ENTPD5 | |
AI196558; AI987697; CD39 antigen-like 4; CD39L4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; Entpd5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; mNTPase; NTPDase 5; NTPDase5; NTPDase-5; Nucleoside diphosphatase; PCPH; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O75356 | |
ENTPD5 | |
Recombinant fragment corresponding to a region within amino acids 1 and 191 of ENTPD5 (Uniprot ID#O75356). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction