Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENTPD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580588
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ENTPD5.
The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.
Specifications
ENTPD5 | |
Polyclonal | |
Unconjugated | |
ENTPD5 | |
AI196558; AI987697; CD39 antigen-like 4; CD39L4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; Entpd5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; mNTPase; NTPDase 5; NTPDase5; NTPDase-5; Nucleoside diphosphatase; PCPH; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
957 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
O75356 | |
ENTPD5 | |
Recombinant Human ENTPD5 protein (Met1-His428). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction