Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTPD5 Recombinant Rabbit Monoclonal Antibody (JE47-87)
GREENER_CHOICE

Catalog No. PIMA534858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534858 Supplier Invitrogen™ Supplier No. MA534858
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Human kidney tissue.

The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE47-87
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene ENTPD5
Gene Accession No. O75356
Gene Alias AI196558; AI987697; CD39 antigen-like 4; CD39L4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; Entpd5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; mNTPase; NTPDase 5; NTPDase5; NTPDase-5; Nucleoside diphosphatase; PCPH; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5
Gene Symbols ENTPD5
Host Species Rabbit
Immunogen Recombinant protein within Human ENTPD5 aa 80-220.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 957
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.