Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTPD6 Monoclonal Antibody (494402)
GREENER_CHOICE

Catalog No. PIMA524201
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524201 Supplier Invitrogen™ Supplier No. MA524201
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blots, no cross-reactivity with recombinant human (rh) CD39, rhCD39L3, or recombinant mouse CD39 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

ENTPD6 is similar to E-type nucleotidases (NTPases). NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD6 contains 4 apyrase-conserved regions which are characteristic of NTPases. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Dec 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD6
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 494402
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Entpd6
Gene Accession No. O75354
Gene Alias 2700026H11Rik; CD39 antigen-like 2; CD39L2; CD39-like 2; dJ738P15.3; Ectonucleoside triphosphate diphosphohydrolase 6; ectonucleoside triphosphate diphosphohydrolase 6 (putative); Entpd6; IL-6SAG; IL6ST2; interleukin 6 signal transducer-2; NTPDase 6; NTPDase-6; RP4-738P15.5
Gene Symbols Entpd6
Host Species Mouse
Immunogen CHO-derived recombinant human CD39L2/ENTPD6 Lys61-Ser484.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 955
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.