Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTPD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584816 Supplier Invitrogen™ Supplier No. PA584816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RMFNRTYKLY SYSYLGLGLM SARLAILGGV EGQPAKDGKE LVSPCLSPSF KGEWEHAEVT YRVSGQKAAA SLHELCAARV SEVLQNRVHR TEEVKHVD Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 82%.

ENTPD6 is similar to E-type nucleotidases (NTPases). NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD6 contains 4 apyrase-conserved regions which are characteristic of NTPases. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Dec 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Entpd6
Gene Accession No. O75354
Gene Alias 2700026H11Rik; CD39 antigen-like 2; CD39L2; CD39-like 2; dJ738P15.3; Ectonucleoside triphosphate diphosphohydrolase 6; ectonucleoside triphosphate diphosphohydrolase 6 (putative); Entpd6; IL-6SAG; IL6ST2; interleukin 6 signal transducer-2; NTPDase 6; NTPDase-6; RP4-738P15.5
Gene Symbols Entpd6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ENTPD6. Recombinant protein control fragment (Product #RP-107698).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.