Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENTPD7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531733
Description
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ENTPD7 is a multi-pass membrane protein. It belongs to the GDA1/CD39 NTPase family. It preferentially hydrolyzes nucleoside 5'-triphosphates. The order of activity with respect to possible substrates is UTP > GTP > CTP.
Specifications
ENTPD7 | |
Polyclonal | |
Unconjugated | |
ENTPD7 | |
1810012B13Rik; 1810020C02Rik; 2810003F23Rik; ectonucleoside triphosphate diphosphohydrolase 7; ENTPD7; Kiaa4066; Lalp1; Lysal2; lysosomal apyrase-like 2; lysosomal apyrase-like protein 1; NTPDase 7; RP11-483F11.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NQZ7 | |
ENTPD7 | |
Recombinant fragment corresponding to a region within amino acids 4 and 217 of Human ENTPD7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction