Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTPD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597710
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597710 Supplier Invitrogen™ Supplier No. PA597710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ENTPD7 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

ENTPD7 is a multi-pass membrane protein. It belongs to the GDA1/CD39 NTPase family. It preferentially hydrolyzes nucleoside 5'-triphosphates. The order of activity with respect to possible substrates is UTP > GTP > CTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ENTPD7
Gene Accession No. Q3TCT4, Q9NQZ7
Gene Alias 1810012B13Rik; 1810020C02Rik; 2810003F23Rik; ectonucleoside triphosphate diphosphohydrolase 7; ENTPD7; Kiaa4066; Lalp1; Lysal2; lysosomal apyrase-like 2; lysosomal apyrase-like protein 1; NTPDase 7; RP11-483F11.1
Gene Symbols ENTPD7
Host Species Rabbit
Immunogen Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 7 protein (51-350AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57089, 93685
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.