Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENTPD7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ENTPD7 |
---|---|
Concentration | 0.48 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4420
![]() ![]() |
Proteintech Group Inc
245581AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ENTPD7 is a multi-pass membrane protein. It belongs to the GDA1/CD39 NTPase family. It preferentially hydrolyzes nucleoside 5′-triphosphates. The order of activity with respect to possible substrates is UTP > GTP > CTP.Specifications
ENTPD7 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9NQZ7 | |
57089 | |
ENTPD7 Fusion Protein Ag18417 | |
Primary | |
-20°C | |
ENTPD7 |
0.48 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ENTPD7, LALP1, NTPDase 7, RP11 483F11.1 | |
ENTPD7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title