Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENTPD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525085
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525085 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525085 Supplier Invitrogen™ Supplier No. PA525085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ENTPD8 Polyclonal Antibody for Western Blot

ENTPD8 is canalicular ectonucleoside NTPDase responsible for the main hepatic NTPDase activity. It is ectonucleoside NTPDases catalyze the hydrolysis of gamma-and beta-phosphate residues of nucleotides, playing a central role in concentration of extracellular nucleotides. ENTPD8 has activity toward ATP, ADP, UTP and UDP, but not toward AMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Entpd8
Gene Accession No. Q5MY95
Gene Alias ecto-ATP-diphosphohydrolase; ectonucleoside triphosphate diphosphohydrolase 8; ENTPD8; E-NTPDase; E-NTPDase 8; GLSR2492; I79_018892; liver ecto-ATP-diphosphohydrolase; NTPDase 8; NTPDase8; NTPDase-8; UNQ2492; UNQ2492/PRO5779
Gene Symbols Entpd8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 396-423 amino acids from the C-terminal region of human ENTPD8.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100755122, 377841
Target Species Human, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.