Learn More
Invitrogen™ ENTPD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113799
Description
Immunogen sequence: IPEAQHRKTP TFLGATAGMR LLSRKNSSQA RDIFAAVTQV LGRSPVDFWG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 79%.
ENTPD8 is canalicular ectonucleoside NTPDase responsible for the main hepatic NTPDase activity. It is ectonucleoside NTPDases catalyze the hydrolysis of gamma-and beta-phosphate residues of nucleotides, playing a central role in concentration of extracellular nucleotides. ENTPD8 has activity toward ATP, ADP, UTP and UDP, but not toward AMP.
Specifications
| ENTPD8 | |
| Polyclonal | |
| Unconjugated | |
| Entpd8 | |
| ecto-ATP-diphosphohydrolase; ectonucleoside triphosphate diphosphohydrolase 8; ENTPD8; E-NTPDase; E-NTPDase 8; GLSR2492; I79_018892; liver ecto-ATP-diphosphohydrolase; NTPDase 8; NTPDase8; NTPDase-8; UNQ2492; UNQ2492/PRO5779 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 377841 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5MY95 | |
| Entpd8 | |
| Synthetic peptide directed towards the N terminal region of human ENTPD8 (aa 110-159). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.