Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENTPD8 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501732261 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2261 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2261 Supplier Proteintech Group Inc Supplier No. 188481AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ENTPD8 is canalicular ectonucleoside NTPDase responsible for the main hepatic NTPDase activity. It is ectonucleoside NTPDases catalyze the hydrolysis of gamma-and beta-phosphate residues of nucleotides, playing a central role in concentration of extracellular nucleotides. ENTPD8 has activity toward ATP, ADP, UTP and UDP, but not toward AMP.

Specifications

Antigen ENTPD8
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ENTPD8
Gene Accession No. Q5MY95
Gene Alias E NTPDase 8, ENTPD8, GLSR2492, NTPDase 8, NTPDase8, UNQ2492
Gene Symbols ENTPD8
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 377841
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.