Learn More
MilliporeSigma™ Supelco™ ENVI™-18 DSK SPE Disk
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Supplier:  MilliporeSigma™ Supelco™ 57170U
 View more versions of this product
Catalog No. 11-100-3694
Only null left
Add to Cart
Description
Description
Retention Mechanism: Reversed-phase. Sample Matrix Compatibility: Aqueous solutions (drinking water). The SPE membrane equivalents of ENVI-18 and ENVI-8 packed bed SPE sorbents. Porous glass fiber membranes embedded with C18 or C8 modified silica particles. Provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples. Typical applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ENVI-18
Food and Beverages
SPE Disk
90 mm
SPE
ENVI
12/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
ENVI is a trademark of Sigma-Aldrich Co. LLC