Learn More
Environmental Express™ Air Monitoring Cassettes
Catalog No. 19057317 Shop All Environmental Express Products
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
19-057317 50/Pk.
1 options
Catalog No. 19-057317 Supplier Environmental Express™ Supplier No. CS25080
Complete line of preloaded air sampling filter cassettes manufactured to meet all applicable NIOSH, OSHA, and EPA standards
  • All cassette housings are manufactured in-house from polystyrene, polypropylene, or conductive materials and are available in two, three, or four piece configurations
  • Cassettes are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads
Specifications
Type PCM Cassettes
Material Polypropylene
Includes Cassette labels
Quantity 50/Pk.