Learn More
Environmental Express™ TCLP Filters
Catalog No. 0973261 Shop All Environmental Express Products
Change view
Click to view available options
Diameter (Metric):
47 mm
90 mm
110 mm
125 mm
142 mm
150 mm
Filter Type:
Prewashed TCLP Filter
Standard TCLP Filter
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Filter Type
09-732-61 110 mm Standard TCLP Filter
09-732-58 150 mm Standard TCLP Filter
09-732-63 47 mm Standard TCLP Filter
09-732-62 90 mm Standard TCLP Filter
09-732-72 47 mm Prewashed TCLP Filter
09-732-76 142 mm Prewashed TCLP Filter
09-732-74 110 mm Prewashed TCLP Filter
09-732-75 125 mm Prewashed TCLP Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 09-732-61 Supplier Environmental Express™ Supplier No. FG75110MM
Add to Cart
Add to Cart
Meets EPA method 1311 requirements.
Meets EPA method 1311 requirements
Borosilicate glass, binder-free filters with 0.7μm pore size. Prepared according to U.S. EPA Method 1311 for TCLP. Come in 47, 90 and 142mm diameters, plus three additional sizes to fit Büchner funnels and Landes zero-headspace extraction units.
Standard TCLP Filters come with Certificate of Analysis warranting that filters meet all guidelines for TCLP extractions and indicating expected background levels of metals.
Prewashed TCLP Filters each receive an acid washing in 1N HNO3, followed by triple rinsing in >1L of deionized water per rinse. Eliminate reagent and waste disposal costs; may reduce blank variability and provide lower levels of background metals compared to onsite washing.
Specifications
Filter Type Standard TCLP Filter
For Use With (Equipment) Landes Zero-headspace Extraction Unit
Material Borosilicate Glass Fiber
For Use With (Application) TCLP extractions and indicating expected background levels of metal
Diameter (English) 4.33 in.
Diameter (Metric) 110 mm