Learn More
3M™ Petrifilm™ Environmental Listeria Plate, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products

Description
- Proven cost effective tool for detection of environmental Listeria
- Provide accurate, efficient, consistent results for food processors and contract labs
- Unique results (qualitative, semi-quantitative and quantitative) allow identification of hot spots in plant, track progress against Listeria over time
- Rapid results in 26 to 30 hours from sample collection
- This plate has been awarded Performance Tested Method (PTM) #030601 for stainless steel, ceramic tile and sealed concrete
Specifications
Specifications
| Content And Storage | Temperatures ≤ 8°C |
| Detects | Listeria |
| Product Type | Count Plate |
| Test Type | Environmental Test |
| For Use With (Application) | Food processors and contract labs |
| Quantity | 200 Pack |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.