Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Petrifilm™ Environmental Listeria Plate, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S30736 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
200 Pack
50 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30736 50 Pack
S30712 200 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S30736 Supplier 3M™ Petrifilm™ Supplier No. 6447
Add to Cart
Add to Cart

Detect the presence of Listeria in environmental samples. A sample ready culture medium containing selective agents, nutrients, a cold water soluble gelling agent and a chromogenic indicator.

  • Proven cost effective tool for detection of environmental Listeria
  • Provide accurate, efficient, consistent results for food processors and contract labs
  • Unique results (qualitative, semi-quantitative and quantitative) allow identification of hot spots in plant, track progress against Listeria over time
  • Rapid results in 26 to 30 hours from sample collection
  • This plate has been awarded Performance Tested Method (PTM) #030601 for stainless steel, ceramic tile and sealed concrete

Specifications

Content And Storage Temperatures ≤ 8°C
Detects Listeria
Product Type Count Plate
Test Type Environmental Test
For Use With (Application) Food processors and contract labs
Quantity 50 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.