Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sonoco™ ThermoSafe Envoy Direct™ Parcel Solutions: 2° to 8°C Coverage
Helps providing cost effective temperature control for regional and national distribution of pharmaceuticals direct from pharmacy. Sonoco™ ThermoSafe Envoy Direct™ Series Parcel Solutions (2° to 8°C Coverage) designed to withstand real-world ambient temperature environments.
$680.00 - $749.50
Specifications
Quantity | 27 Shippers/Cs. |
---|---|
Width (English) Exterior | 8.88 in. |
Includes | Foam shipper with outercarton, corrugate payload box and foam buffer pads |
Type | Parcel Solution |
Temperature Range (Metric) | 2° to 8°C |
Catalog Number | Mfr. No. | Payload | Dimensions (LxWxH) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Payload | Dimensions (LxWxH) | Price | Quantity & Availability | ||||
03-600-182
|
Sonoco™
ED4R1 |
3.7 L | Payload: 8.88 x 8.88 x 2.88 in. |
Case of 27 for $680.00
|
|
||||
03-600-184
|
Sonoco™
ED5R1 |
5.6 L | Payload: 10.38 x 8.88 x 3.38 in. |
Case of 27 for $749.50
|
|
||||
Description
- 2° to 8°C measured throughout the entire product space
- Ideal for next day delivery
- Simple and fast assembly drives operational efficiencies
- Uses minimal components with seasonal conditioning requirements to simplify inventory management
- Optimized dimensional weight reduces transportation costs
Specifications
27 Shippers/Cs. | |
Foam shipper with outercarton, corrugate payload box and foam buffer pads | |
2° to 8°C |
8.88 in. | |
Parcel Solution |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title