Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Envy™ Foaming Disinfectant Cleaner

Catalog No. 17000038
Click to view available options
Volume (Metric):
539 g

Foaming, quaternary-based, hospital-use disinfectant cleaner that cleans, disinfects and deodorizes in one step.

  • Quat based disinfectant cleaner
  • Foaming action speeds cleaning performance
  • Convenient aerosol
  • Foam clings to vertical surfaces
  • Clean lavender fragrance

Specifications

Product Type Quat-based Disinfectant
pH 12.1
Sterility Non-sterile
Appearance White
Container Type Aerosol
Dilution Ready to use. Do not dilute.
For Use With (Application) Use on hard, nonporous surfaces including vinyl, painted surfaces, plastic, glazed ceramic, glazed porcelain, chrome, laminated surfaces and baked enamel surfaces; Effective on stains in health care, educational, lodging and food service environments
Quantity 12 Pk.
Volume (English) 19 oz.
Volume (Metric) 539 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.