Learn More
Invitrogen™ ENY2 Monoclonal Antibody (GT245)

Description
Positive Control: U87-MG, SK-N-SH, IMR32 Predicted Reactivity: Mouse (100%), Zebrafish (85%), Xenopus laevis (93%), Pig (100%), Chicken (96%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ENY2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT245 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | ENY2 |
| Gene Accession No. | Q9NPA8 |
| Gene Alias | 1810057B09Rik; 6720481I12; DC6; e(y)2; e(y)2 homolog; e(y)2 protein; enhancer of yellow 2 homolog; enhancer of yellow 2 homolog (Drosophila); enhancer of yellow 2 transcription factor homolog; enhancer of yellow eye homolog; ENY 2; ENY2; ENY-2; ENY2, transcription and export complex 2 subunit; Ey2; Sus1; transcription and mRNA export factor ENY2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.