Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Enzootic Bovine Leukosis Virus Ab ELISA Kit

Catalog No. 89032339
Click to view available options
Quantity:
1 Kit

Enzyme-linked immunosorbent assay for the detection of antibodies to Enzootic Bovine Leukosis Virus (EBLV)

  • Used for the in vitro quantitative determination of human SOD1 in human serum, human plasma, cell lysate, culture supernatants and buffered solution
  • Assay will recognize native and recombinant human IL-9

Specifications

Description Enzootic Bovine Leukosis Virus Ab ELISA Kit is an enzyme-linked immunosorbent assay for the detection of antibodies to Enzootic Bovine Leukosis Virus (EBLV).
Conjugate HRP
Detection Method Colorimetric
Product Type Enzootic Bovine Leukosis Virus Ab ELISA Kit
Sample Type Serum
Target Species Bovine
For Use With (Application) Qualitative
Kit Contents Microtitre Plate: 96 wells, Negative Control Serum: 10x (0.4mL), Positive Control Serum - limit: 10x (0.4mL), Conjugate: 100x (0.8mL), Sample Diluent 13 (240mL), Conjugate Diluent 11 (70mL), TMB-Complete 5 (60mL), Wash Solution: 20x (2 x 60mL), Stop Solution: Acid solution ready to use (60mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 10 μL (Prediluted)
Storage Requirements Store the kit at 4°C. Do not freeze.
Target Bovine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.