Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enzymatic Digest Casein Hydrolysate

Catalog No. p-7076796
Click to view available options
Quantity:
11.3 kg (25 lb.)
2.2 kg (5 lb.)
453.5 g (1 lb.)

Enzymatic digest of casein.

  • Synonym: Peptones
  • pH: 6.6 to 7.5
  • CAS: 65072-00-6
  • MDLNumber: MFCD00282824
TRUSTED_SUSTAINABILITY

Specifications

Name Casein hydrolysate
pH Range 6.6 to 7.5
Quantity 453.5 g (1 lb.)
Packaging Plastic bottle
Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.