Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Enzymes and the Process of Digestion
GREENER_CHOICESCI_ED

Catalog No. S05800 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05800 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer.
Catalog No. S05800 Supplier Innovating Science™ Supplier No. IS3009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer.

Understanding the vital role that enzymes play in the process of human digestive

  • All the food in the world is of no use if the human body does not have the ability to extract necessary nutrients from it
  • With this activity, students expose three nutrients (carbohydrates, proteins and lipids) to different digestive enzymes
  • These samples tested against nutrients to which no enzymes are added to determine if the enzymes were effective in digesting the compounds
  • Upon completion, students understand the importance of the digestive system and the vital role enzymes play in releasing nutrients from food and converting them to a form usable by the body
  • Contains enough materials for 15 groups.
  • Kit Includes:
    • Pancreatin (Powder, 5 g)
    • Bile Salts (Powder, 0.25 g)
    • Albumin Egg (Powder, 2.5 g)
    • Soluble Starch (2.5 g)
    • Pepsin 1:10,000 (1 g)
    • Amylase Bacteriological Powder (25 g)
    • 0.1M Sodium hydroxide (25 mL)
    • 0.1M Hydrochloric acid (25 mL)
    • Biuret for protein test (25 mL)
    • 1% Phenolphthalein (25 mL)
    • Dilute lugols solution (25 mL)
    • Pure Olive Oil (25 mL)
    • 10 Plastic Pipets

Compliance

DOT: Small quantity exemption 173.3. Package conforms to 49CFR 173.4.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Learning about the vital role that enzymes play in the human digestive process
Quantity 1 Kit
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Enzymes and the Process of Digestion Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.