Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EO 1428
GREENER_CHOICESDP

Catalog No. 290850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-085-0 50 mg
29-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-085-0 Supplier R&D Systems™ Supplier No. 2908/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of p38α and p38β2

Selective inhibitor of p38α and p38β2 that displays no activity at p38γ, p38δ, ERK1/2 and JNK1. Inhibits production of inflammatory cytokines including IL-8, TNF-α, IL-6, IL-1β and IL-10 (IC50 values are 4, 5, 17, 30 and 74 nM respectively). Displays anti-inflammatory activity in murine models of acute and chronic dermatitis.

Chemical Identifiers

CAS 321351-00-2
Molecular Formula C20H16BrClN2O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 415.71
Purity >98%
Molecular Weight (g/mol) 415.715
Chemical Name or Material EO 1428
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.