Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EOGT/AER61 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305987MFV610
Description
EOGT/AER61 Polyclonal antibody specifically detects EOGT/AER61 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
EOGT/AER61 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
AER61, AER61 glycosyltransferase, chromosome 3 open reading frame 64, EC 2.4, FLJ13078, FLJ33770, FLJ41219, MGC34132 | |
Produced in rabbits immunized with E. coli-derived Human EOGT/AER61 fragment. | |
0.1 mL | |
Proteases & Other Enzymes | |
285203 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction